The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194286.3 | NP_919262.2 | 13 - 13 | 8425 | 94 |
GenBank mRNA | AK055717.1 | - | 1 - 1 | 1861 | 94 |
AK125400.1 | - | 1 - 1 | 3164 | 94 | |
BM680369.1 | - | 1 - 1 | 94 |