Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL5A1
Entrez Gene ID
1289
Gene Name
collagen type V alpha 1
Gene Aliases
EDSC
Chromosome Location
Chr.9: 134641790 - 134844843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000093.4 NP_000084.3 40 - 41 3589 83
NM_001278074.1 NP_001265003.1 40 - 41 3589 83
XM_017014266.1 XP_016869755.1 40 - 41 3604 83
GenBank mRNA AB209864.1 - 39 - 40 3068 83
AB371583.1 - 40 - 41 3589 83
D90279.1 - 40 - 41 3332 83

View Full Product Details