The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000093.4 | NP_000084.3 | 59 - 60 | 4992 | 75 |
NM_001278074.1 | NP_001265003.1 | 59 - 60 | 4992 | 75 | |
XM_017014266.1 | XP_016869755.1 | 59 - 60 | 5007 | 75 | |
GenBank mRNA | AB209864.1 | - | 58 - 59 | 4471 | 75 |
AB371583.1 | - | 59 - 60 | 4992 | 75 | |
BC008760.2 | - | 12 - 13 | 868 | 75 | |
D90279.1 | - | 59 - 60 | 4735 | 75 |