The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000477.5 | NP_000468.1 | 12 - 13 | 1723 | 141 |
GenBank mRNA | A06977.1 | - | 12 - 13 | 1725 | 141 |
AF190168.1 | - | 12 - 13 | 1650 | 141 | |
AF542069.1 | - | 12 - 13 | 1683 | 141 | |
AK292755.1 | - | 12 - 13 | 1688 | 141 | |
AK298437.1 | - | 12 - 13 | 1716 | 141 | |
AK298461.1 | - | 12 - 13 | 1433 | 141 | |
AK314794.1 | - | 12 - 13 | 1691 | 141 | |
AY544124.1 | - | 8 - 9 | 1081 | 141 | |
AY550967.1 | - | 12 - 13 | 1657 | 141 | |
AY728024.1 | - | 12 - 13 | 1656 | 141 | |
AY960291.1 | - | 12 - 13 | 1682 | 141 | |
BC014308.1 | - | 3 - 4 | 428 | 141 | |
BC034023.1 | - | 12 - 13 | 1691 | 141 | |
BC035969.1 | - | 8 - 9 | 1052 | 141 | |
BC036003.1 | - | 12 - 13 | 1691 | 141 | |
BC041789.1 | - | 8 - 9 | 1115 | 141 | |
DQ986150.1 | - | 12 - 13 | 1661 | 141 | |
HQ537426.1 | - | 11 - 12 | 1578 | 141 | |
V00494.1 | - | 12 - 13 | 1689 | 141 | |
V00495.1 | - | 12 - 13 | 1725 | 141 |