Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA1B
Entrez Gene ID
774
Gene Name
calcium voltage-gated channel subunit alpha1 B
Gene Aliases
BIII, CACNL1A5, CACNN, Cav2.2, DYT23
Chromosome Location
Chr.9: 137877789 - 138124624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000718.3 NP_000709.1 19 - 20 3214 55
NM_001243812.1 NP_001230741.1 19 - 20 3214 55
XM_011518991.2 XP_011517293.1 19 - 20 3181 55
XM_011518992.2 XP_011517294.1 19 - 20 3178 55
XM_011518993.2 XP_011517295.1 19 - 20 3177 55
XM_011518994.2 XP_011517296.1 19 - 20 3180 55
XM_017015115.1 XP_016870604.1 19 - 20 3178 55
XM_017015116.1 XP_016870605.1 19 - 20 3181 55
XM_017015117.1 XP_016870606.1 19 - 20 3180 55
XM_017015118.1 XP_016870607.1 19 - 20 3174 55
XM_017015119.1 XP_016870608.1 19 - 20 3180 55
XM_017015120.1 XP_016870609.1 19 - 20 3177 55
GenBank mRNA AB209467.1 - 1 - 2 158 55
M94172.1 - 19 - 20 3214 55
M94173.1 - 19 - 20 3214 55

View Full Product Details