Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE9A
Entrez Gene ID
5152
Gene Name
phosphodiesterase 9A
Gene Aliases
HSPDE9A2
Chromosome Location
Chr.21: 42653752 - 42775509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001567.1 NP_001001567.1 16 - 17 1471 69
NM_001001568.1 NP_001001568.1 14 - 15 1322 69
NM_001001569.1 NP_001001569.1 13 - 14 1267 69
NM_001001570.1 NP_001001570.1 16 - 17 1492 69
NM_001001571.1 NP_001001571.1 15 - 16 1400 69
NM_001001572.1 NP_001001572.1 13 - 14 1278 69
NM_001001573.1 NP_001001573.1 14 - 15 1458 69
NM_001001574.1 NP_001001574.1 15 - 16 1393 69
NM_001001575.1 NP_001001575.1 12 - 13 1223 69
NM_001001576.1 NP_001001576.1 12 - 13 1182 69
NM_001001577.1 NP_001001577.1 13 - 14 1301 69
NM_001001578.1 NP_001001578.1 14 - 15 1345 69
NM_001001579.1 NP_001001579.1 14 - 15 1338 69
NM_001001580.1 NP_001001580.1 11 - 12 1138 69
NM_001001581.1 NP_001001581.1 15 - 16 1502 69
NM_001001582.1 NP_001001582.1 16 - 17 1580 69
NM_001001583.1 NP_001001583.1 16 - 17 1573 69
NM_001001584.2 NP_001001584.1 15 - 16 1401 69
NM_001001585.1 NP_001001585.1 15 - 16 1525 69
NM_001315533.1 NP_001302462.1 17 - 18 1547 69
NM_002606.2 NP_002597.1 17 - 18 1651 69
XM_011529598.2 XP_011527900.1 17 - 18 1573 69
XM_017028366.1 XP_016883855.1 15 - 16 1389 69
GenBank mRNA AF048837.1 - 17 - 18 1648 69
AF067223.1 - 17 - 18 1600 69
AF067224.1 - 16 - 17 1419 69
AF067225.1 - 14 - 15 1261 69
AF067226.1 - 13 - 14 1206 69
AK001165.1 - 14 - 15 1253 69
AK001307.1 - 16 - 17 1442 69
AK127770.1 - 7 - 8 3002 69
AK294112.1 - 14 - 15 1288 69
AK314679.1 - 17 - 18 1624 69
AY196299.1 - 16 - 17 1465 69
AY196300.1 - 15 - 16 1373 69
AY196301.1 - 13 - 14 1251 69
AY196302.1 - 14 - 15 1431 69
AY196303.1 - 15 - 16 1366 69
AY196304.1 - 12 - 13 1196 69
AY196305.1 - 12 - 13 1155 69
AY196306.1 - 13 - 14 1274 69
AY196307.1 - 14 - 15 1318 69
AY196308.1 - 14 - 15 1311 69
AY196309.1 - 11 - 12 1111 69
AY196310.1 - 15 - 16 1475 69
AY196311.1 - 16 - 17 1553 69
AY196312.1 - 16 - 17 1546 69
AY196313.1 - 15 - 16 1373 69
AY196314.1 - 15 - 16 1498 69
AY242121.1 - 15 - 16 1337 69
AY701187.1 - 17 - 18 1544 69
BC009047.1 - 16 - 17 1445 69
BT007016.1 - 16 - 17 1408 69
KU178246.1 - 16 - 17 1408 69
KU178247.1 - 17 - 18 1588 69
KU178248.1 - 13 - 14 1204 69

View Full Product Details