Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE9A
Entrez Gene ID
5152
Gene Name
phosphodiesterase 9A
Gene Aliases
HSPDE9A2
Chromosome Location
Chr.21: 42653752 - 42775509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001581.1 NP_001001581.1 2 - 3 178 62
NM_001001582.1 NP_001001582.1 3 - 4 256 62
NM_001001583.1 NP_001001583.1 3 - 4 249 62
NM_001001585.1 NP_001001585.1 3 - 4 256 62
NM_002606.2 NP_002597.1 4 - 5 327 62
XM_011529598.2 XP_011527900.1 4 - 5 249 62
XM_011529600.2 XP_011527902.1 4 - 5 300 62
XM_017028367.1 XP_016883856.1 4 - 5 300 62
GenBank mRNA AF048837.1 - 4 - 5 324 62
AF067223.1 - 4 - 5 276 62
AK314679.1 - 4 - 5 300 62
AY196310.1 - 2 - 3 151 62
AY196311.1 - 3 - 4 229 62
AY196312.1 - 3 - 4 222 62
AY196314.1 - 3 - 4 229 62
KU178247.1 - 4 - 5 264 62

View Full Product Details