Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE9A
Entrez Gene ID
5152
Gene Name
phosphodiesterase 9A
Gene Aliases
HSPDE9A2
Chromosome Location
Chr.21: 42653752 - 42775509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001567.1 NP_001001567.1 8 - 9 619 59
NM_001001568.1 NP_001001568.1 6 - 7 470 59
NM_001001569.1 NP_001001569.1 5 - 6 415 59
NM_001001570.1 NP_001001570.1 8 - 9 640 59
NM_001001571.1 NP_001001571.1 7 - 8 548 59
NM_001001572.1 NP_001001572.1 5 - 6 426 59
NM_001001573.1 NP_001001573.1 6 - 7 606 59
NM_001001574.1 NP_001001574.1 7 - 8 541 59
NM_001001575.1 NP_001001575.1 4 - 5 371 59
NM_001001576.1 NP_001001576.1 4 - 5 330 59
NM_001001577.1 NP_001001577.1 5 - 6 449 59
NM_001001578.1 NP_001001578.1 6 - 7 493 59
NM_001001579.1 NP_001001579.1 6 - 7 486 59
NM_001001580.1 NP_001001580.1 3 - 4 286 59
NM_001001581.1 NP_001001581.1 7 - 8 650 59
NM_001001582.1 NP_001001582.1 8 - 9 728 59
NM_001001583.1 NP_001001583.1 8 - 9 721 59
NM_001001584.2 NP_001001584.1 7 - 8 549 59
NM_001001585.1 NP_001001585.1 7 - 8 673 59
NM_001315533.1 NP_001302462.1 9 - 10 695 59
NM_002606.2 NP_002597.1 9 - 10 799 59
XM_011529598.2 XP_011527900.1 9 - 10 721 59
XM_011529600.2 XP_011527902.1 9 - 10 772 59
XM_017028366.1 XP_016883855.1 7 - 8 537 59
XM_017028367.1 XP_016883856.1 9 - 10 772 59
GenBank mRNA AF048837.1 - 9 - 10 796 59
AF067223.1 - 9 - 10 748 59
AF067224.1 - 8 - 9 567 59
AF067225.1 - 6 - 7 409 59
AF067226.1 - 5 - 6 354 59
AK001165.1 - 6 - 7 401 59
AK001307.1 - 8 - 9 590 59
AK294112.1 - 6 - 7 436 59
AK314679.1 - 9 - 10 772 59
AY196299.1 - 8 - 9 613 59
AY196300.1 - 7 - 8 521 59
AY196301.1 - 5 - 6 399 59
AY196302.1 - 6 - 7 579 59
AY196303.1 - 7 - 8 514 59
AY196304.1 - 4 - 5 344 59
AY196305.1 - 4 - 5 303 59
AY196306.1 - 5 - 6 422 59
AY196307.1 - 6 - 7 466 59
AY196308.1 - 6 - 7 459 59
AY196309.1 - 3 - 4 259 59
AY196310.1 - 7 - 8 623 59
AY196311.1 - 8 - 9 701 59
AY196312.1 - 8 - 9 694 59
AY196313.1 - 7 - 8 521 59
AY196314.1 - 7 - 8 646 59
AY242121.1 - 7 - 8 485 59
AY701187.1 - 9 - 10 692 59
BC009047.1 - 8 - 9 593 59
BT007016.1 - 8 - 9 556 59
KU178246.1 - 8 - 9 556 59
KU178247.1 - 9 - 10 736 59
KU178248.1 - 5 - 6 352 59

View Full Product Details