Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRB14
Entrez Gene ID
2888
Gene Name
growth factor receptor bound protein 14
Gene Aliases
-
Chromosome Location
Chr.2: 164492406 - 164621850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303422.1 NP_001290351.1 4 - 5 591 130
NM_004490.2 NP_004481.2 5 - 6 1220 130
XM_011511022.2 XP_011509324.1 5 - 6 852 130
XM_017003899.1 XP_016859388.1 5 - 6 657 130
GenBank mRNA AK301961.1 - 4 - 5 536 130
AK302129.1 - 4 - 5 548 130
AK307744.1 - 5 - 6 732 130
AK309590.1 - 5 - 6 658 130
AK313153.1 - 5 - 6 849 130
BC053559.1 - 5 - 6 690 130
L76687.1 - 5 - 6 1219 130

View Full Product Details