The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004655.3 | NP_004646.3 | 2 - 3 | 1104 | 95 |
XM_011525319.2 | XP_011523621.1 | 2 - 3 | 1315 | 95 | |
XM_011525320.1 | XP_011523622.1 | 2 - 3 | 1304 | 95 | |
XM_011525321.2 | XP_011523623.1 | 2 - 3 | 1252 | 95 | |
XM_017025192.1 | XP_016880681.1 | 2 - 3 | 971 | 95 | |
XM_017025193.1 | XP_016880682.1 | 2 - 3 | 1304 | 95 | |
XM_017025194.1 | XP_016880683.1 | 2 - 3 | 1083 | 95 | |
GenBank mRNA | AF078165.1 | - | 1 - 2 | 904 | 95 |
AF205888.1 | - | 1 - 2 | 921 | 95 | |
AK025718.1 | - | 1 - 2 | 126 | 95 | |
AK315303.1 | - | 2 - 3 | 1020 | 95 | |
BC101533.1 | - | 1 - 2 | 884 | 95 | |
BC143244.1 | - | 1 - 2 | 884 | 95 | |
CR933657.1 | - | 2 - 3 | 1129 | 95 |