The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047160.2 | NP_001040625.1 | 10 - 11 | 1336 | 106 |
NM_005863.4 | NP_005854.2 | 8 - 9 | 1247 | 106 | |
NR_073040.1 | 8 - 9 | 1280 | 106 | ||
GenBank mRNA | AB209852.1 | - | 10 - 11 | 1282 | 106 |
AJ010046.1 | - | 8 - 9 | 1180 | 106 | |
AK024919.1 | - | 8 - 9 | 1187 | 106 | |
AK294244.1 | - | 4 - 5 | 713 | 106 | |
AK299335.1 | - | 10 - 11 | 1292 | 106 | |
AK302687.1 | - | 10 - 11 | 1118 | 106 | |
AK304028.1 | - | 8 - 9 | 1222 | 106 | |
AK315566.1 | - | 8 - 9 | 1189 | 106 | |
BC010285.1 | - | 8 - 9 | 1178 | 106 | |
BC053553.1 | - | 10 - 11 | 1288 | 106 | |
BX537509.1 | - | 4 - 5 | 3885 | 106 | |
U02081.1 | - | 6 - 7 | 768 | 106 |