The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006083.3 | NP_006074.2 | 8 - 9 | 745 | 111 |
GenBank mRNA | AF182645.1 | - | 8 - 9 | 746 | 111 |
AJ005579.1 | - | 8 - 9 | 635 | 111 | |
AK225924.1 | - | 8 - 9 | 729 | 111 | |
AK293626.1 | - | 8 - 9 | 573 | 111 | |
AK308730.1 | - | 8 - 9 | 583 | 111 | |
AK312363.1 | - | 8 - 9 | 715 | 111 | |
AY007094.1 | - | 8 - 9 | 669 | 111 | |
BC013005.2 | - | 8 - 9 | 700 | 111 | |
BC051295.1 | - | 8 - 9 | 703 | 111 | |
BC071964.1 | - | 8 - 9 | 693 | 111 |