Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBTF
Entrez Gene ID
7343
Gene Name
upstream binding transcription factor, RNA polymerase I
Gene Aliases
NOR-90, UBF, UBF-1, UBF1, UBF2
Chromosome Location
Chr.17: 44205033 - 44221626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076683.1 NP_001070151.1 14 - 15 1723 113
NM_001076684.2 NP_001070152.1 14 - 15 1809 113
NM_014233.3 NP_055048.1 15 - 16 1920 113
NR_045058.1 15 - 16 2117 113
XM_006722059.3 XP_006722122.1 15 - 16 1959 113
XM_006722060.2 XP_006722123.1 14 - 15 1703 113
XM_006722061.2 XP_006722124.1 15 - 16 1787 113
XM_017024995.1 XP_016880484.1 15 - 16 1968 113
XM_017024996.1 XP_016880485.1 15 - 16 1920 113
XM_017024997.1 XP_016880486.1 15 - 16 2112 113
XM_017024998.1 XP_016880487.1 15 - 16 1828 113
XM_017024999.1 XP_016880488.1 14 - 15 1703 113
XM_017025000.1 XP_016880489.1 14 - 15 1761 113
XM_017025001.1 XP_016880490.1 14 - 15 1809 113
XM_017025002.1 XP_016880491.1 14 - 15 1689 113
XM_017025003.1 XP_016880492.1 14 - 15 1699 113
XM_017025004.1 XP_016880493.1 14 - 15 1691 113
GenBank mRNA AK291733.1 - 15 - 16 1745 113
AK292518.1 - 15 - 16 2117 113
AK292577.1 - 15 - 16 2005 113
AK296917.1 - 9 - 10 1033 113
AK298008.1 - 12 - 13 1335 113
AK308321.1 - 13 - 14 1785 113
BC042297.1 - 14 - 15 1714 113
X53390.1 - 14 - 15 1771 113
X53461.1 - 14 - 15 1771 113
X56687.1 - 14 - 15 1692 113

View Full Product Details