Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UBTF
- Entrez Gene ID
-
7343
- Gene Name
- upstream binding transcription factor, RNA polymerase I
- Gene Aliases
-
NOR-90, UBF, UBF-1, UBF1, UBF2
- Chromosome Location
-
Chr.17: 44205033 - 44221626 on Build GRCh38
View Full Product Details