The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018558.3 | NP_061028.3 | 4 - 5 | 543 | 69 |
XM_011531184.2 | XP_011529486.1 | 4 - 5 | 831 | 69 | |
GenBank mRNA | AF144648.1 | - | 4 - 5 | 508 | 69 |
AF189259.1 | - | 4 - 5 | 543 | 69 | |
BC109210.1 | - | 4 - 5 | 543 | 69 | |
BC109211.1 | - | 4 - 5 | 543 | 69 |