Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196983856-196984129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR5
Entrez Gene ID
81494
Gene Name
complement factor H related 5
Gene Aliases
CFHL5, CFHR5D, FHR-5, FHR5
UniGene
Hs.282594
Gene Chromosome Location
Chr.1: 196975022-197009725 on Build GRCh38
Transcripts
NM_030787.3 XM_011510020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1724743 c.455C>T p.S152L Chr.1: 196984090-196984090 on Build GRCh38
6339468 c.377T>A p.L126Q Chr.1: 196984012-196984012 on Build GRCh38
5379018 c.254-1G>A p.? Chr.1: 196983960-196983960 on Build GRCh38
4026554 c.372G>T p.E124D Chr.1: 196984079-196984079 on Build GRCh38
7135723 c.430C>A p.L144I Chr.1: 196984065-196984065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746505923 Chr.1: 196984104-196984104 on Build GRCh38
rs766096486 Chr.1: 196984019-196984019 on Build GRCh38
rs181626516 Chr.1: 196983908-196983908 on Build GRCh38
rs754934306 Chr.1: 196984033-196984033 on Build GRCh38
rs774353375 Chr.1: 196984090-196984090 on Build GRCh38
rs746609986 Chr.1: 196983928-196983928 on Build GRCh38
rs746240586 Chr.1: 196984074-196984074 on Build GRCh38
rs756576112 Chr.1: 196984050-196984050 on Build GRCh38
rs758371568 Chr.1: 196984101-196984101 on Build GRCh38
rs552606505 Chr.1: 196983893-196983893 on Build GRCh38
rs886595279 Chr.1: 196983968-196983968 on Build GRCh38
rs973112968 Chr.1: 196983884-196983884 on Build GRCh38
rs114023763 Chr.1: 196983953-196983953 on Build GRCh38
rs1054274655 Chr.1: 196983982-196983982 on Build GRCh38
rs770559120 Chr.1: 196984064-196984064 on Build GRCh38
rs962387786 Chr.1: 196984015-196984015 on Build GRCh38
rs747654308 Chr.1: 196983915-196983915 on Build GRCh38
rs770245831 Chr.1: 196984077-196984077 on Build GRCh38
rs375328527 Chr.1: 196984056-196984056 on Build GRCh38
rs966940369 Chr.1: 196984096-196984096 on Build GRCh38
rs546578427 Chr.1: 196983929-196983929 on Build GRCh38
rs757464725 Chr.1: 196984017-196984017 on Build GRCh38
rs147278469 Chr.1: 196984009-196984009 on Build GRCh38
rs147791058 Chr.1: 196984091-196984091 on Build GRCh38
rs764351908 Chr.1: 196983992-196983992 on Build GRCh38
rs912772838 Chr.1: 196983919-196983919 on Build GRCh38
rs1039021834 Chr.1: 196983914-196983914 on Build GRCh38
rs141053543 Chr.1: 196984092-196984092 on Build GRCh38
rs190122254 Chr.1: 196983963-196983963 on Build GRCh38
rs775616599 Chr.1: 196983965-196983965 on Build GRCh38
rs762992026 Chr.1: 196983990-196983990 on Build GRCh38
rs781568341 Chr.1: 196983950-196983951 on Build GRCh38
rs771889857 Chr.1: 196983923-196983923 on Build GRCh38
rs368988109 Chr.1: 196984083-196984083 on Build GRCh38
rs919046647 Chr.1: 196983898-196983898 on Build GRCh38
rs777378052 Chr.1: 196984039-196984039 on Build GRCh38
rs202069780 Chr.1: 196983912-196983913 on Build GRCh38
rs185709089 Chr.1: 196983956-196983956 on Build GRCh38
rs570596954 Chr.1: 196983895-196983895 on Build GRCh38
rs576196434 Chr.1: 196983955-196983955 on Build GRCh38
rs777447274 Chr.1: 196984102-196984102 on Build GRCh38
rs778342266 Chr.1: 196983911-196983911 on Build GRCh38
rs971025311 Chr.1: 196984054-196984054 on Build GRCh38
rs764194328 Chr.1: 196984086-196984086 on Build GRCh38
rs756898801 Chr.1: 196984105-196984105 on Build GRCh38
rs772976239 Chr.1: 196983927-196983927 on Build GRCh38
rs536828292 Chr.1: 196984034-196984034 on Build GRCh38
rs367909871 Chr.1: 196983938-196983938 on Build GRCh38
rs751805375 Chr.1: 196984002-196984002 on Build GRCh38
rs1016469143 Chr.1: 196984011-196984011 on Build GRCh38
rs753650925 Chr.1: 196984029-196984029 on Build GRCh38
rs752687157 Chr.1: 196984097-196984097 on Build GRCh38
rs775956342 Chr.1: 196984082-196984082 on Build GRCh38
rs61745675 Chr.1: 196984037-196984037 on Build GRCh38
rs318240754 Chr.1: 196984021-196984021 on Build GRCh38
rs778029757 Chr.1: 196984068-196984068 on Build GRCh38
rs140691305 Chr.1: 196984036-196984036 on Build GRCh38
rs748590126 Chr.1: 196983983-196983984 on Build GRCh38
rs754791022 Chr.1: 196984093-196984093 on Build GRCh38

View Full Product Details