Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24092728-24092997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
UniGene
Hs.460355
Gene Chromosome Location
Chr.16: 23835979-24220611 on Build GRCh38
Transcripts
NM_002738.6 NM_212535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5435647 c.530-8T>G p.? Chr.16: 24092783-24092783 on Build GRCh38
3507912 c.668G>A p.W223* Chr.16: 24092929-24092929 on Build GRCh38
3507906 c.610C>T p.P204S Chr.16: 24092871-24092871 on Build GRCh38
4059460 c.532A>C p.R178R Chr.16: 24092793-24092793 on Build GRCh38
5700728 c.611C>A p.P204H Chr.16: 24092872-24092872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747105951 Chr.16: 24092847-24092847 on Build GRCh38
rs1029252769 Chr.16: 24092794-24092794 on Build GRCh38
rs917317004 Chr.16: 24092894-24092894 on Build GRCh38
rs3729896 Chr.16: 24092867-24092867 on Build GRCh38
rs759210165 Chr.16: 24092782-24092782 on Build GRCh38
rs543897172 Chr.16: 24092846-24092846 on Build GRCh38
rs773891890 Chr.16: 24092901-24092901 on Build GRCh38
rs754963571 Chr.16: 24092759-24092759 on Build GRCh38
rs779094238 Chr.16: 24092796-24092796 on Build GRCh38
rs748510627 Chr.16: 24092862-24092862 on Build GRCh38
rs202027145 Chr.16: 24092964-24092964 on Build GRCh38
rs762695587 Chr.16: 24092897-24092897 on Build GRCh38
rs755974100 Chr.16: 24092783-24092783 on Build GRCh38
rs573366163 Chr.16: 24092811-24092811 on Build GRCh38
rs751409221 Chr.16: 24092755-24092755 on Build GRCh38
rs372031319 Chr.16: 24092975-24092975 on Build GRCh38
rs769253047 Chr.16: 24092849-24092849 on Build GRCh38
rs777343122 Chr.16: 24092855-24092855 on Build GRCh38
rs767494458 Chr.16: 24092771-24092771 on Build GRCh38
rs758345369 Chr.16: 24092832-24092832 on Build GRCh38
rs746115990 Chr.16: 24092810-24092810 on Build GRCh38
rs1005322850 Chr.16: 24092813-24092813 on Build GRCh38
rs752461659 Chr.16: 24092772-24092772 on Build GRCh38
rs759011396 Chr.16: 24092953-24092953 on Build GRCh38
rs199950702 Chr.16: 24092967-24092967 on Build GRCh38
rs770291872 Chr.16: 24092863-24092863 on Build GRCh38
rs752290250 Chr.16: 24092958-24092958 on Build GRCh38
rs781543122 Chr.16: 24092950-24092950 on Build GRCh38

View Full Product Details