Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43587287-43587518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLH
Entrez Gene ID
5429
Gene Name
DNA polymerase eta
Gene Aliases
RAD30, RAD30A, XP-V, XPV
UniGene
Hs.655467
Gene Chromosome Location
Chr.6: 43576141-43620523 on Build GRCh38
Transcripts
NM_001291969.1 NM_001291970.1 NM_006502.2 XM_005249186.3 XM_011514698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3777698 c.475G>C p.E159Q Chr.6: 43587474-43587474 on Build GRCh38
1672447 c.334G>A p.A112T Chr.6: 43587333-43587333 on Build GRCh38
13899 c.458G>A p.G153D Chr.6: 43587457-43587457 on Build GRCh38
4382703 c.332G>A p.R111H Chr.6: 43587331-43587331 on Build GRCh38
1079643 c.373G>T p.V125L Chr.6: 43587372-43587372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926622952 Chr.6: 43587477-43587477 on Build GRCh38
rs121908563 Chr.6: 43587375-43587375 on Build GRCh38
rs759875951 Chr.6: 43587355-43587355 on Build GRCh38
rs749395920 Chr.6: 43587467-43587467 on Build GRCh38
rs138298839 Chr.6: 43587354-43587354 on Build GRCh38
rs570093755 Chr.6: 43587339-43587339 on Build GRCh38
rs146061840 Chr.6: 43587436-43587436 on Build GRCh38
rs747435372 Chr.6: 43587445-43587445 on Build GRCh38
rs758423288 Chr.6: 43587331-43587331 on Build GRCh38
rs56239418 Chr.6: 43587329-43587329 on Build GRCh38
rs772706799 Chr.6: 43587325-43587325 on Build GRCh38
rs755826058 Chr.6: 43587413-43587413 on Build GRCh38
rs142827517 Chr.6: 43587412-43587412 on Build GRCh38
rs537497046 Chr.6: 43587347-43587347 on Build GRCh38
rs375000061 Chr.6: 43587330-43587330 on Build GRCh38
rs2307464 Chr.6: 43587446-43587446 on Build GRCh38
rs768567392 Chr.6: 43587478-43587478 on Build GRCh38
rs367709714 Chr.6: 43587457-43587457 on Build GRCh38
rs775769629 Chr.6: 43587374-43587374 on Build GRCh38
rs767433001 Chr.6: 43587489-43587489 on Build GRCh38
rs766019609 Chr.6: 43587419-43587419 on Build GRCh38
rs774374609 Chr.6: 43587479-43587479 on Build GRCh38
rs370592983 Chr.6: 43587318-43587318 on Build GRCh38
rs781680603 Chr.6: 43587435-43587436 on Build GRCh38
rs773061849 Chr.6: 43587494-43587494 on Build GRCh38
rs370505776 Chr.6: 43587386-43587386 on Build GRCh38
rs1031362847 Chr.6: 43587432-43587432 on Build GRCh38
rs554936509 Chr.6: 43587414-43587414 on Build GRCh38
rs200366966 Chr.6: 43587439-43587439 on Build GRCh38
rs755734466 Chr.6: 43587313-43587313 on Build GRCh38
rs767257383 Chr.6: 43587408-43587408 on Build GRCh38
rs745988408 Chr.6: 43587492-43587492 on Build GRCh38
rs781354148 Chr.6: 43587367-43587367 on Build GRCh38
rs761707571 Chr.6: 43587488-43587488 on Build GRCh38
rs761688287 Chr.6: 43587402-43587402 on Build GRCh38
rs762937711 Chr.6: 43587381-43587381 on Build GRCh38
rs753397463 Chr.6: 43587428-43587428 on Build GRCh38
rs748739006 Chr.6: 43587319-43587319 on Build GRCh38
rs997248275 Chr.6: 43587423-43587423 on Build GRCh38
rs781723095 Chr.6: 43587452-43587452 on Build GRCh38
rs139742736 Chr.6: 43587466-43587466 on Build GRCh38
rs144449059 Chr.6: 43587465-43587465 on Build GRCh38
rs201090378 Chr.6: 43587370-43587370 on Build GRCh38
rs764092379 Chr.6: 43587384-43587384 on Build GRCh38

View Full Product Details