Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31269153-31269422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-C
Entrez Gene ID
3107
Gene Name
major histocompatibility complex, class I, C
Gene Aliases
D6S204, HLA-JY3, HLAC, HLC-C, MHC, PSORS1
UniGene
Hs.656020
Gene Chromosome Location
Chr.6: 31268749-31272136 on Build GRCh38
Transcripts
NM_001243042.1 NM_002117.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4160664 c.1049G>C p.C350S Chr.6: 31269385-31269385 on Build GRCh38
6173670 c.1186G>A p.E396K Chr.6: 31269359-31269359 on Build GRCh38
6173671 c.1093G>A p.E365K Chr.6: 31269359-31269359 on Build GRCh38
5991810 c.1083delC p.I362fs*>5 Chr.6: 31269351-31269351 on Build GRCh38
4160661 c.1087A>G p.T363A Chr.6: 31269347-31269347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527971789 Chr.6: 31269208-31269208 on Build GRCh38
rs984616135 Chr.6: 31269371-31269371 on Build GRCh38
rs17885557 Chr.6: 31269310-31269310 on Build GRCh38
rs3998381 Chr.6: 31269237-31269237 on Build GRCh38
rs773199105 Chr.6: 31269319-31269319 on Build GRCh38
rs770655323 Chr.6: 31269400-31269400 on Build GRCh38
rs763576150 Chr.6: 31269348-31269348 on Build GRCh38
rs113599490 Chr.6: 31269258-31269258 on Build GRCh38
rs776254781 Chr.6: 31269337-31269337 on Build GRCh38
rs375542879 Chr.6: 31269327-31269327 on Build GRCh38
rs765974199 Chr.6: 31269189-31269189 on Build GRCh38
rs765733803 Chr.6: 31269343-31269343 on Build GRCh38
rs758717642 Chr.6: 31269293-31269293 on Build GRCh38
rs777991288 Chr.6: 31269397-31269398 on Build GRCh38
rs192019825 Chr.6: 31269182-31269182 on Build GRCh38
rs746845038 Chr.6: 31269399-31269399 on Build GRCh38
rs778748999 Chr.6: 31269309-31269309 on Build GRCh38
rs759559219 Chr.6: 31269331-31269331 on Build GRCh38
rs113668674 Chr.6: 31269176-31269176 on Build GRCh38
rs9264594 Chr.6: 31269271-31269271 on Build GRCh38
rs386698954 Chr.6: 31269237-31269239 on Build GRCh38
rs761537700 Chr.6: 31269177-31269177 on Build GRCh38
rs41544716 Chr.6: 31269339-31269339 on Build GRCh38
rs767160115 Chr.6: 31269180-31269180 on Build GRCh38
rs1055337554 Chr.6: 31269391-31269391 on Build GRCh38
rs781221565 Chr.6: 31269303-31269303 on Build GRCh38
rs753054241 Chr.6: 31269288-31269288 on Build GRCh38
rs41555414 Chr.6: 31269263-31269263 on Build GRCh38
rs111924491 Chr.6: 31269251-31269251 on Build GRCh38
rs2001181 Chr.6: 31269221-31269221 on Build GRCh38
rs772202592 Chr.6: 31269392-31269392 on Build GRCh38
rs2394961 Chr.6: 31269250-31269250 on Build GRCh38
rs371039627 Chr.6: 31269308-31269308 on Build GRCh38
rs41543813 Chr.6: 31269229-31269229 on Build GRCh38
rs386698955 Chr.6: 31269237-31269240 on Build GRCh38
rs879017580 Chr.6: 31269321-31269321 on Build GRCh38
rs751964800 Chr.6: 31269354-31269354 on Build GRCh38
rs765288380 Chr.6: 31269182-31269183 on Build GRCh38
rs907750566 Chr.6: 31269394-31269394 on Build GRCh38
rs41544314 Chr.6: 31269239-31269239 on Build GRCh38
rs745997118 Chr.6: 31269307-31269307 on Build GRCh38
rs1130838 Chr.6: 31269347-31269347 on Build GRCh38
rs35708511 Chr.6: 31269385-31269385 on Build GRCh38
rs750476831 Chr.6: 31269377-31269377 on Build GRCh38
rs112392852 Chr.6: 31269244-31269244 on Build GRCh38
rs757448813 Chr.6: 31269359-31269359 on Build GRCh38
rs281860591 Chr.6: 31269398-31269398 on Build GRCh38
rs372648063 Chr.6: 31269194-31269194 on Build GRCh38
rs2394960 Chr.6: 31269240-31269240 on Build GRCh38
rs41548117 Chr.6: 31269324-31269324 on Build GRCh38
rs757545357 Chr.6: 31269302-31269302 on Build GRCh38
rs781416483 Chr.6: 31269376-31269376 on Build GRCh38
rs751770408 Chr.6: 31269298-31269298 on Build GRCh38
rs760399331 Chr.6: 31269324-31269324 on Build GRCh38
rs1000145319 Chr.6: 31269294-31269294 on Build GRCh38
rs777918128 Chr.6: 31269295-31269295 on Build GRCh38
rs111260491 Chr.6: 31269275-31269275 on Build GRCh38
rs2894204 Chr.6: 31269284-31269284 on Build GRCh38
rs758988637 Chr.6: 31269338-31269338 on Build GRCh38
rs760203049 Chr.6: 31269187-31269187 on Build GRCh38
rs922217117 Chr.6: 31269281-31269281 on Build GRCh38
rs895081373 Chr.6: 31269402-31269402 on Build GRCh38
rs1013600389 Chr.6: 31269233-31269233 on Build GRCh38
rs17885436 Chr.6: 31269225-31269225 on Build GRCh38
rs61759946 Chr.6: 31269382-31269382 on Build GRCh38

View Full Product Details