Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24112808-24113081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
UniGene
Hs.460355
Gene Chromosome Location
Chr.16: 23835979-24220611 on Build GRCh38
Transcripts
NM_002738.6 NM_212535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1222021 c.911A>C p.K304T Chr.16: 24113062-24113062 on Build GRCh38
379882 c.907C>A p.Q303K Chr.16: 24113058-24113058 on Build GRCh38
3507919 c.834G>A p.L278L Chr.16: 24112985-24112985 on Build GRCh38
5038833 c.905_906insT p.Q303fs*4 Chr.16: 24113056-24113057 on Build GRCh38
5805410 c.871C>A p.P291T Chr.16: 24113022-24113022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779177002 Chr.16: 24112964-24112964 on Build GRCh38
rs768632460 Chr.16: 24112928-24112928 on Build GRCh38
rs199901715 Chr.16: 24113032-24113032 on Build GRCh38
rs778332947 Chr.16: 24112951-24112951 on Build GRCh38
rs375302170 Chr.16: 24112934-24112934 on Build GRCh38
rs566935963 Chr.16: 24112896-24112896 on Build GRCh38
rs11268093 Chr.16: 24112843-24112843 on Build GRCh38
rs879713249 Chr.16: 24112952-24112952 on Build GRCh38
rs761610718 Chr.16: 24112932-24112932 on Build GRCh38
rs745475837 Chr.16: 24112978-24112978 on Build GRCh38
rs756332028 Chr.16: 24112950-24112950 on Build GRCh38
rs373653332 Chr.16: 24112843-24112852 on Build GRCh38
rs17847884 Chr.16: 24113000-24113000 on Build GRCh38
rs568306326 Chr.16: 24113024-24113024 on Build GRCh38
rs200159126 Chr.16: 24112838-24112838 on Build GRCh38
rs934334806 Chr.16: 24112847-24112847 on Build GRCh38
rs536939564 Chr.16: 24113055-24113055 on Build GRCh38
rs762879227 Chr.16: 24113056-24113056 on Build GRCh38
rs68132728 Chr.16: 24112829-24112838 on Build GRCh38
rs779746680 Chr.16: 24113015-24113015 on Build GRCh38
rs115598469 Chr.16: 24113057-24113057 on Build GRCh38
rs998179536 Chr.16: 24113034-24113034 on Build GRCh38
rs748062707 Chr.16: 24113039-24113039 on Build GRCh38
rs533390919 Chr.16: 24112840-24112840 on Build GRCh38
rs369360721 Chr.16: 24112937-24112937 on Build GRCh38
rs377353224 Chr.16: 24112867-24112867 on Build GRCh38
rs776488997 Chr.16: 24112929-24112929 on Build GRCh38
rs1044347262 Chr.16: 24112883-24112883 on Build GRCh38
rs752791417 Chr.16: 24112940-24112940 on Build GRCh38
rs746506850 Chr.16: 24113021-24113021 on Build GRCh38
rs992929483 Chr.16: 24112859-24112859 on Build GRCh38
rs182283658 Chr.16: 24112933-24112933 on Build GRCh38
rs11646540 Chr.16: 24112863-24112863 on Build GRCh38
rs747698479 Chr.16: 24112928-24112932 on Build GRCh38
rs1005300794 Chr.16: 24112889-24112889 on Build GRCh38
rs3831621 Chr.16: 24112845-24112854 on Build GRCh38
rs373144544 Chr.16: 24112955-24112955 on Build GRCh38
rs147975795 Chr.16: 24113044-24113044 on Build GRCh38

View Full Product Details