Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28404770-28405041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC46A1
Entrez Gene ID
113235
Gene Name
solute carrier family 46 member 1
Gene Aliases
G21, HCP1, PCFT
UniGene
Hs.446689
Gene Chromosome Location
Chr.17: 28394642-28406630 on Build GRCh38
Transcripts
NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4064899 c.775G>A p.A259T Chr.17: 28404922-28404922 on Build GRCh38
6782534 c.909A>C p.L303L Chr.17: 28404788-28404788 on Build GRCh38
6582855 c.754G>T p.V252F Chr.17: 28404943-28404943 on Build GRCh38
6782536 c.727T>G p.F243V Chr.17: 28404970-28404970 on Build GRCh38
4129769 c.926C>A p.A309E Chr.17: 28404771-28404771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781978069 Chr.17: 28404880-28404880 on Build GRCh38
rs782790090 Chr.17: 28404887-28404887 on Build GRCh38
rs782773007 Chr.17: 28404912-28404913 on Build GRCh38
rs558367155 Chr.17: 28404840-28404840 on Build GRCh38
rs886052755 Chr.17: 28404793-28404793 on Build GRCh38
rs781828817 Chr.17: 28404923-28404923 on Build GRCh38
rs782194324 Chr.17: 28404830-28404830 on Build GRCh38
rs782387872 Chr.17: 28404872-28404872 on Build GRCh38
rs782009707 Chr.17: 28404808-28404808 on Build GRCh38
rs376504291 Chr.17: 28404820-28404820 on Build GRCh38
rs201540617 Chr.17: 28404799-28404799 on Build GRCh38
rs782306668 Chr.17: 28404848-28404848 on Build GRCh38
rs782240737 Chr.17: 28404965-28404965 on Build GRCh38
rs782571400 Chr.17: 28404958-28404958 on Build GRCh38
rs782253488 Chr.17: 28404813-28404813 on Build GRCh38
rs370955621 Chr.17: 28404922-28404922 on Build GRCh38
rs34552966 Chr.17: 28404814-28404814 on Build GRCh38
rs782013829 Chr.17: 28404966-28404966 on Build GRCh38
rs369228677 Chr.17: 28404804-28404804 on Build GRCh38
rs782077797 Chr.17: 28404861-28404861 on Build GRCh38
rs185900287 Chr.17: 28404941-28404941 on Build GRCh38
rs782651103 Chr.17: 28404971-28404971 on Build GRCh38
rs782222724 Chr.17: 28404960-28404960 on Build GRCh38
rs782060856 Chr.17: 28404968-28404968 on Build GRCh38
rs781906324 Chr.17: 28405010-28405010 on Build GRCh38
rs782161981 Chr.17: 28404883-28404883 on Build GRCh38
rs374413378 Chr.17: 28404933-28404933 on Build GRCh38
rs781800160 Chr.17: 28404888-28404888 on Build GRCh38
rs782729175 Chr.17: 28404913-28404913 on Build GRCh38
rs782105362 Chr.17: 28404905-28404905 on Build GRCh38
rs781968978 Chr.17: 28404854-28404854 on Build GRCh38
rs782457383 Chr.17: 28404945-28404945 on Build GRCh38
rs41297081 Chr.17: 28404967-28404967 on Build GRCh38
rs782781428 Chr.17: 28404986-28404986 on Build GRCh38
rs782820497 Chr.17: 28404976-28404976 on Build GRCh38

View Full Product Details