The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003338.4 | NP_003329.1 | 2 - 3 | 314 | 84 |
XM_017016609.1 | XP_016872098.1 | 2 - 3 | 213 | 84 | |
GenBank mRNA | AF257220.1 | - | 2 - 3 | 237 | 84 |
AJ272367.1 | - | 2 - 3 | 237 | 84 | |
AK291901.1 | - | 2 - 3 | 285 | 84 | |
BC005980.1 | - | 2 - 3 | 285 | 84 | |
BC015997.1 | - | 2 - 3 | 289 | 84 | |
BI598342.1 | - | 2 - 3 | 318 | 84 | |
BT007041.1 | - | 2 - 3 | 91 | 84 | |
DA677507.1 | - | 2 - 3 | 314 | 84 | |
X78140.1 | - | 2 - 3 | 106 | 84 |