The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020799.3 | NP_065850.1 | 9 - 10 | 1659 | 65 |
XM_006717928.2 | XP_006717991.1 | 10 - 11 | 1760 | 65 | |
XM_006717930.3 | XP_006717993.1 | 9 - 10 | 2247 | 65 | |
XM_011539983.1 | XP_011538285.1 | 10 - 11 | 2167 | 65 | |
XM_011539985.1 | XP_011538287.1 | 10 - 11 | 1490 | 65 | |
XM_017016453.1 | XP_016871942.1 | 10 - 11 | 2484 | 65 | |
XM_017016454.1 | XP_016871943.1 | 11 - 12 | 1968 | 65 | |
XM_017016455.1 | XP_016871944.1 | 12 - 13 | 2423 | 65 | |
XM_017016456.1 | XP_016871945.1 | 10 - 11 | 2080 | 65 | |
XM_017016457.1 | XP_016871946.1 | 8 - 9 | 1868 | 65 | |
XM_017016458.1 | XP_016871947.1 | 7 - 8 | 1184 | 65 | |
GenBank mRNA | AB010120.1 | - | 8 - 9 | 1187 | 65 |
AB037794.1 | - | 9 - 10 | 1980 | 65 | |
AK056086.1 | - | 9 - 10 | 1588 | 65 | |
AK090678.1 | - | 4 - 5 | 1480 | 65 | |
AK092946.1 | - | 8 - 9 | 1788 | 65 | |
BC010846.1 | - | 9 - 10 | 1577 | 65 | |
BX649096.1 | - | 10 - 11 | 10082 | 65 |