Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIX5
Entrez Gene ID
147912
Gene Name
SIX homeobox 5
Gene Aliases
BOR2, DMAHP
Chromosome Location
Chr.19: 45764785 - 45769239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC033204.1 - 1 - 2 223 66

View Full Product Details