The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178452.5 | NP_848547.4 | 2 - 3 | 441 | 105 |
XM_006721129.2 | XP_006721192.1 | 2 - 3 | 441 | 105 | |
XM_011522853.2 | XP_011521155.1 | 2 - 3 | 441 | 105 | |
XM_011522854.2 | XP_011521156.1 | 2 - 3 | 441 | 105 | |
XM_011522855.2 | XP_011521157.1 | 2 - 3 | 441 | 105 | |
XM_011522857.2 | XP_011521159.1 | 2 - 3 | 441 | 105 | |
XM_011522858.2 | XP_011521160.1 | 2 - 3 | 441 | 105 | |
XM_017022918.1 | XP_016878407.1 | 2 - 3 | 441 | 105 | |
XM_017022919.1 | XP_016878408.1 | 2 - 3 | 749 | 105 | |
GenBank mRNA | AK057238.1 | - | 2 - 3 | 393 | 105 |
AK310961.1 | - | 2 - 3 | 392 | 105 | |
AL833327.1 | - | 2 - 3 | 412 | 105 | |
AL833328.1 | - | 2 - 3 | 412 | 105 | |
AL833336.1 | - | 2 - 3 | 412 | 105 | |
BC024009.2 | - | 2 - 3 | 384 | 105 | |
BX648657.1 | - | 2 - 3 | 414 | 105 | |
DA349893.1 | - | 2 - 3 | 447 | 105 |