The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178527.3 | NP_848622.2 | 19 - 20 | 2789 | 69 |
XM_011509426.1 | XP_011507728.1 | 19 - 20 | 2789 | 69 | |
XM_017001065.1 | XP_016856554.1 | 19 - 20 | 2484 | 69 | |
XM_017001066.1 | XP_016856555.1 | 19 - 20 | 2478 | 69 | |
XM_017001067.1 | XP_016856556.1 | 18 - 19 | 2669 | 69 | |
XM_017001068.1 | XP_016856557.1 | 18 - 19 | 2688 | 69 | |
XM_017001069.1 | XP_016856558.1 | 18 - 19 | 2383 | 69 | |
XM_017001070.1 | XP_016856559.1 | 18 - 19 | 2377 | 69 | |
XM_017001071.1 | XP_016856560.1 | 19 - 20 | 2396 | 69 | |
XM_017001072.1 | XP_016856561.1 | 17 - 18 | 2522 | 69 | |
XM_017001073.1 | XP_016856562.1 | 18 - 19 | 2295 | 69 | |
XM_017001074.1 | XP_016856563.1 | 17 - 18 | 2224 | 69 | |
XM_017001075.1 | XP_016856564.1 | 12 - 13 | 1551 | 69 | |
XM_017001076.1 | XP_016856565.1 | 9 - 10 | 1158 | 69 | |
GenBank mRNA | AK128104.1 | - | 19 - 20 | 2789 | 69 |
AK302731.1 | - | 16 - 17 | 2186 | 69 | |
BC042592.1 | - | 19 - 20 | 2736 | 69 | |
BX648119.1 | - | 12 - 13 | 1700 | 69 |