Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD42
Entrez Gene ID
338699
Gene Name
ankyrin repeat domain 42
Gene Aliases
PPP1R79, SARP
Chromosome Location
Chr.11: 83193739 - 83256099 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300972.1 NP_001287901.1 7 - 8 1850 121
NM_001300973.1 NP_001287902.1 7 - 8 1847 121
NM_001300975.1 NP_001287904.1 7 - 8 1850 121
NM_182603.3 NP_872409.2 7 - 8 1766 121
GenBank mRNA AK095193.1 - 7 - 8 996 121
BC062729.1 - 1 - 2 139 121
BC171752.1 - 7 - 8 944 121
DQ508815.1 - 7 - 8 1191 121
EF041819.1 - 7 - 8 1710 121

View Full Product Details