The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182632.2 | NP_872438.2 | 4 - 5 | 744 | 78 |
GenBank mRNA | AK055798.1 | - | 4 - 5 | 744 | 78 |
AK298257.1 | - | 4 - 5 | 646 | 78 | |
BC056757.1 | - | 4 - 5 | 702 | 78 |