Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS9
Entrez Gene ID
64965
Gene Name
mitochondrial ribosomal protein S9
Gene Aliases
MRP-S9, RPMS9, S9mt
Chromosome Location
Chr.2: 105038025 - 105099960 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182640.2 NP_872578.1 5 - 6 552 113
XM_011511644.2 XP_011509946.1 4 - 5 392 113
GenBank mRNA AK130078.1 - 5 - 6 493 113
BC036575.1 - 6 - 7 505 113
BC047784.1 - 4 - 5 202 113
BC057240.1 - 5 - 6 506 113

View Full Product Details