The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182658.2 | NP_872599.1 | 11 - 12 | 1308 | 62 |
GenBank mRNA | AF549189.1 | - | 11 - 12 | 1215 | 62 |
BC113398.1 | - | 11 - 12 | 1308 | 62 | |
BC113400.1 | - | 11 - 12 | 1308 | 62 |