The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015420.6 | NP_056235.4 | 1 - 1 | 652 | 96 |
GenBank mRNA | AK027799.1 | - | 1 - 1 | 653 | 96 |
AK074725.1 | - | 1 - 1 | 617 | 96 | |
BC101810.1 | - | 1 - 1 | 442 | 96 | |
BC112042.1 | - | 1 - 1 | 442 | 96 |