The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182528.3 | NP_872334.2 | 2 - 2 | 1960 | 64 |
GenBank mRNA | AF525315.1 | - | 2 - 2 | 1959 | 64 |