Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAT4
Entrez Gene ID
79633
Gene Name
FAT atypical cadherin 4
Gene Aliases
CDHF14, CDHR11, FAT-J, FATJ, HKLLS2, NBLA00548, VMLDS2
Chromosome Location
Chr.4: 125314913 - 125492932 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291285.1 NP_001278214.1 17 - 17 14275 65
NM_001291303.1 NP_001278232.1 17 - 17 14278 65
NM_024582.4 NP_078858.4 17 - 17 14272 65
XM_011532236.2 XP_011530538.1 18 - 18 15355 65
XM_011532237.2 XP_011530539.1 17 - 17 10163 65
GenBank mRNA AK026709.1 - 1 - 1 354 65
AK091292.1 - 1 - 1 922 65
AK096646.1 - 9 - 9 2571 65
AK291461.1 - 9 - 9 2815 65
AY356402.1 - 15 - 15 8995 65

View Full Product Details