Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39864847-39865084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAAM2
Entrez Gene ID
23500
Gene Name
dishevelled associated activator of morphogenesis 2
Gene Aliases
dJ90A20A.1
UniGene
Hs.357128
Gene Chromosome Location
Chr.6: 39792366-39904877 on Build GRCh38
Transcripts
NM_001201427.1 NM_015345.3 XM_006715039.3 XM_006715040.3 XM_006715042.2 XM_006715043.2 XM_006715045.3 XM_006715046.3 XM_017010630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3627052 c.391G>A p.E131K Chr.6: 39865037-39865037 on Build GRCh38
5915517 c.370G>A p.E124K Chr.6: 39865016-39865016 on Build GRCh38
6547227 c.361G>T p.E121* Chr.6: 39865007-39865007 on Build GRCh38
7400703 c.355G>C p.D119H Chr.6: 39865001-39865001 on Build GRCh38
6639687 c.348C>T p.Y116Y Chr.6: 39864994-39864994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944593386 Chr.6: 39864972-39864972 on Build GRCh38
rs754592396 Chr.6: 39864980-39864980 on Build GRCh38
rs199678029 Chr.6: 39864973-39864973 on Build GRCh38
rs760457649 Chr.6: 39864963-39864963 on Build GRCh38
rs775187998 Chr.6: 39864919-39864919 on Build GRCh38
rs573533468 Chr.6: 39864993-39864993 on Build GRCh38
rs535422672 Chr.6: 39864873-39864873 on Build GRCh38
rs1010407216 Chr.6: 39864896-39864896 on Build GRCh38
rs746263836 Chr.6: 39864997-39864997 on Build GRCh38
rs377713135 Chr.6: 39864939-39864939 on Build GRCh38
rs770301199 Chr.6: 39865000-39865000 on Build GRCh38
rs543612445 Chr.6: 39865059-39865059 on Build GRCh38
rs371082714 Chr.6: 39865014-39865014 on Build GRCh38
rs757931255 Chr.6: 39864942-39864942 on Build GRCh38
rs766276356 Chr.6: 39864930-39864930 on Build GRCh38
rs761984417 Chr.6: 39865037-39865037 on Build GRCh38
rs867024285 Chr.6: 39864964-39864964 on Build GRCh38
rs553776054 Chr.6: 39864894-39864894 on Build GRCh38
rs1054533824 Chr.6: 39864870-39864870 on Build GRCh38
rs749360733 Chr.6: 39865015-39865015 on Build GRCh38
rs907461153 Chr.6: 39864991-39864991 on Build GRCh38
rs747735397 Chr.6: 39864995-39864995 on Build GRCh38
rs574596379 Chr.6: 39864996-39864996 on Build GRCh38
rs554462066 Chr.6: 39864906-39864906 on Build GRCh38
rs774422861 Chr.6: 39865033-39865033 on Build GRCh38
rs778576748 Chr.6: 39864983-39864983 on Build GRCh38
rs375635373 Chr.6: 39865043-39865043 on Build GRCh38
rs776885499 Chr.6: 39864975-39864975 on Build GRCh38

View Full Product Details