Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43600895-43601166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLH
Entrez Gene ID
5429
Gene Name
DNA polymerase eta
Gene Aliases
RAD30, RAD30A, XP-V, XPV
UniGene
Hs.655467
Gene Chromosome Location
Chr.6: 43576141-43620523 on Build GRCh38
Transcripts
NM_001291969.1 NM_001291970.1 NM_006502.2 XM_005249186.3 XM_011514698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6513981 c.741C>A p.F247L Chr.6: 43601068-43601068 on Build GRCh38
7308535 c.700C>T p.R234C Chr.6: 43601027-43601027 on Build GRCh38
6304689 c.758G>A p.R253H Chr.6: 43601085-43601085 on Build GRCh38
1741760 c.757C>T p.R253C Chr.6: 43601084-43601084 on Build GRCh38
7293545 c.754A>G p.I252V Chr.6: 43601081-43601081 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs2307460 Chr.6: 43601005-43601005 on Build GRCh38
rs772778835 Chr.6: 43600999-43601000 on Build GRCh38
rs781009488 Chr.6: 43601050-43601050 on Build GRCh38
rs771570891 Chr.6: 43601090-43601090 on Build GRCh38
rs769582858 Chr.6: 43600963-43600963 on Build GRCh38
rs571127888 Chr.6: 43601068-43601068 on Build GRCh38
rs199869019 Chr.6: 43600945-43600945 on Build GRCh38
rs202063884 Chr.6: 43601062-43601062 on Build GRCh38
rs745778317 Chr.6: 43601052-43601052 on Build GRCh38
rs770838265 Chr.6: 43600944-43600944 on Build GRCh38
rs61756403 Chr.6: 43601025-43601025 on Build GRCh38
rs760141371 Chr.6: 43601110-43601110 on Build GRCh38
rs765722828 Chr.6: 43601111-43601111 on Build GRCh38
rs763234525 Chr.6: 43601132-43601132 on Build GRCh38
rs562876596 Chr.6: 43600946-43600946 on Build GRCh38
rs750486517 Chr.6: 43601133-43601133 on Build GRCh38
rs780041698 Chr.6: 43601063-43601063 on Build GRCh38
rs559506859 Chr.6: 43600988-43600988 on Build GRCh38
rs934271402 Chr.6: 43601131-43601131 on Build GRCh38
rs777882970 Chr.6: 43600940-43600940 on Build GRCh38
rs774074015 Chr.6: 43600996-43600996 on Build GRCh38
rs764294319 Chr.6: 43601027-43601027 on Build GRCh38
rs766810281 Chr.6: 43601000-43601000 on Build GRCh38
rs77588969 Chr.6: 43601020-43601020 on Build GRCh38
rs771582888 Chr.6: 43600971-43600972 on Build GRCh38
rs751639745 Chr.6: 43601042-43601042 on Build GRCh38
rs775902831 Chr.6: 43601112-43601112 on Build GRCh38
rs765677072 Chr.6: 43601006-43601006 on Build GRCh38
rs777147317 Chr.6: 43601001-43601001 on Build GRCh38
rs772570523 Chr.6: 43601092-43601092 on Build GRCh38
rs538518658 Chr.6: 43601084-43601084 on Build GRCh38
rs145530456 Chr.6: 43601065-43601065 on Build GRCh38
rs747076276 Chr.6: 43600943-43600943 on Build GRCh38
rs755973003 Chr.6: 43601056-43601056 on Build GRCh38
rs373892236 Chr.6: 43601077-43601077 on Build GRCh38
rs761322378 Chr.6: 43600998-43600998 on Build GRCh38
rs866837387 Chr.6: 43601078-43601078 on Build GRCh38
rs548691073 Chr.6: 43601007-43601007 on Build GRCh38
rs370185041 Chr.6: 43601046-43601046 on Build GRCh38
rs376072346 Chr.6: 43600978-43600978 on Build GRCh38
rs992033293 Chr.6: 43600987-43600987 on Build GRCh38

View Full Product Details