The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322831.1 | NP_001309760.1 | 20 - 21 | 2686 | 94 |
NM_001322834.1 | NP_001309763.1 | 20 - 21 | 2686 | 94 | |
NM_031913.4 | NP_114119.2 | 20 - 21 | 2686 | 94 | |
NR_136409.1 | 20 - 21 | 2920 | 94 | ||
XM_017007307.1 | XP_016862796.1 | 19 - 20 | 2138 | 94 | |
GenBank mRNA | AJ697972.1 | - | 2 - 3 | 543 | 94 |
BC037292.1 | - | 19 - 20 | 3644 | 94 | |
BQ435892.1 | - | 2 - 3 | 416 | 94 | |
DQ993202.1 | - | 20 - 21 | 2460 | 94 |