The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017520.3 | NP_059990.2 | 8 - 9 | 2031 | 64 |
XM_011535124.2 | XP_011533426.1 | 8 - 9 | 2032 | 64 | |
GenBank mRNA | AJ293409.1 | - | 3 - 4 | 334 | 64 |
AK056785.1 | - | 1 - 2 | 78 | 64 | |
AK300258.1 | - | 8 - 9 | 2031 | 64 | |
AL832864.1 | - | 8 - 9 | 1980 | 64 | |
BC003542.1 | - | 8 - 9 | 1939 | 64 | |
BC046214.1 | - | 2 - 3 | 218 | 64 |