The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040710.2 | NP_001035800.1 | 5 - 6 | 546 | 102 |
GenBank mRNA | BC050462.1 | - | 5 - 6 | 486 | 102 |