Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPOP
Entrez Gene ID
8405
Gene Name
speckle type BTB/POZ protein
Gene Aliases
BTBD32, TEF2
Chromosome Location
Chr.17: 49598884 - 49678163 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007226.1 NP_001007227.1 4 - 5 547 65
NM_001007227.1 NP_001007228.1 3 - 4 425 65
NM_001007228.1 NP_001007229.1 2 - 3 373 65
NM_001007229.1 NP_001007230.1 3 - 4 424 65
NM_001007230.1 NP_001007231.1 4 - 5 476 65
NM_003563.3 NP_003554.1 3 - 4 495 65
XM_005257723.4 XP_005257780.1 3 - 4 719 65
XM_005257724.4 XP_005257781.1 2 - 3 649 65
XM_017025204.1 XP_016880693.1 1 - 2 162 65
GenBank mRNA AJ000644.1 - 2 - 3 233 65
AK222589.1 - 2 - 3 198 65
AK293045.1 - 3 - 4 706 65
AK293587.1 - 2 - 3 315 65
AK312691.1 - 2 - 3 209 65
BC001269.1 - 3 - 4 379 65
BC003385.1 - 2 - 3 190 65
CB992730.1 - 4 - 5 448 65

View Full Product Details