Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
-
Chromosome Location
Chr.17: 44141902 - 44162477 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024032.4 NP_076937.2 5 - 6 1691 105
XM_011525187.1 XP_011523489.1 5 - 6 1538 105
XM_011525188.1 XP_011523490.1 4 - 5 1457 105
XM_011525191.2 XP_011523493.1 5 - 6 1541 105
XM_011525192.1 XP_011523494.1 5 - 6 1541 105
XM_011525194.1 XP_011523496.1 5 - 6 1541 105
XM_011525197.2 XP_011523499.1 2 - 3 706 105
XM_017025031.1 XP_016880520.1 5 - 6 1714 105
GenBank mRNA AL834295.1 - 5 - 6 1635 105

View Full Product Details