Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
-
Chromosome Location
Chr.17: 44141902 - 44162477 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171251.2 NP_001164722.1 6 - 7 1799 72
NM_001321310.1 NP_001308239.1 4 - 5 1574 72
NM_001321311.1 NP_001308240.1 6 - 7 1796 72
NM_024032.4 NP_076937.2 6 - 7 1802 72
XM_011525187.1 XP_011523489.1 6 - 7 1649 72
XM_011525188.1 XP_011523490.1 5 - 6 1568 72
XM_011525189.1 XP_011523491.1 5 - 6 1511 72
XM_011525190.1 XP_011523492.1 5 - 6 1508 72
XM_011525191.2 XP_011523493.1 6 - 7 1652 72
XM_011525193.1 XP_011523495.1 4 - 5 1424 72
XM_011525197.2 XP_011523499.1 3 - 4 817 72
XM_017025031.1 XP_016880520.1 6 - 7 1825 72
XM_017025032.1 XP_016880521.1 6 - 7 1821 72
GenBank mRNA AK291924.1 - 6 - 7 1793 72
AL834295.1 - 6 - 7 1746 72
BC000121.1 - 4 - 5 1454 72

View Full Product Details