Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15orf57
Entrez Gene ID
90416
Gene Name
chromosome 15 open reading frame 57
Gene Aliases
CCDC32
Chromosome Location
Chr.15: 40531341 - 40565057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080791.2 NP_001074260.1 3 - 4 704 69
NM_001080792.2 NP_001074261.1 3 - 4 494 69
NM_052849.3 NP_443081.1 3 - 4 679 69
GenBank mRNA AK092214.1 - 3 - 4 627 69
AK312842.1 - 3 - 4 469 69
AL832032.2 - 2 - 3 426 69
BC001673.2 - 2 - 3 418 69
BG709516.1 - 3 - 4 497 69
BP202606.1 - 3 - 4 471 69
BQ878848.1 - 3 - 4 453 69
BU657334.1 - 3 - 4 468 69
BX404085.2 - 3 - 4 673 69
CD686743.1 - 1 - 2 139 69

View Full Product Details