The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181833.2 | NP_861971.1 | 4 - 5 | 892 | 69 |
GenBank mRNA | AF122827.1 | - | 4 - 5 | 499 | 69 |