Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJB3
Entrez Gene ID
414061
Gene Name
DnaJ heat shock protein family (Hsp40) member B3
Gene Aliases
HCG3
Chromosome Location
Chr.2: 233742750 - 233744015 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001394.3 NP_001001394.1 1 - 1 248 160
GenBank mRNA AY083665.1 - 1 - 1 248 160
BC017590.2 - 1 - 1 248 160
BC024013.2 - 1 - 1 211 160

View Full Product Details