The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020179.2 | NP_064564.1 | 3 - 3 | 766 | 139 |
XM_011542907.1 | XP_011541209.1 | 3 - 3 | 791 | 139 | |
XM_011542908.2 | XP_011541210.1 | 4 - 4 | 894 | 139 | |
XM_011542909.1 | XP_011541211.1 | 3 - 3 | 776 | 139 | |
XM_011542910.2 | XP_011541212.1 | 3 - 3 | 1529 | 139 | |
XM_011542911.2 | XP_011541213.1 | 3 - 3 | 1455 | 139 | |
XM_017018019.1 | XP_016873508.1 | 3 - 3 | 1511 | 139 | |
XM_017018020.1 | XP_016873509.1 | 4 - 4 | 952 | 139 | |
GenBank mRNA | AF197137.1 | - | 3 - 3 | 740 | 139 |
BC031564.1 | - | 3 - 3 | 760 | 133 |