Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WASF2
Entrez Gene ID
10163
Gene Name
WAS protein family member 2
Gene Aliases
IMD2, SCAR2, WASF4, WAVE2, dJ393P12.2
Chromosome Location
Chr.1: 27404226 - 27490187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201404.2 NP_001188333.1 1 - 1 88 112
NM_006990.4 NP_008921.1 1 - 1 88 112
GenBank mRNA BC012329.1 - 1 - 1 59 112
BC040943.1 - 1 - 1 50 112
DC376823.1 - 1 - 1 71 112
HY001891.1 - 1 - 1 68 112

View Full Product Details