The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024306.4 | NP_077282.3 | 2 - 3 | 441 | 84 |
XM_011523317.2 | XP_011521619.1 | 2 - 3 | 433 | 84 | |
XM_011523319.2 | XP_011521621.1 | 2 - 3 | 140 | 84 | |
GenBank mRNA | AJ278219.1 | - | 1 - 2 | 89 | 84 |
AK058016.1 | - | 2 - 3 | 435 | 84 | |
AK315512.1 | - | 2 - 3 | 254 | 84 | |
BC002679.2 | - | 2 - 3 | 407 | 84 | |
BC004263.1 | - | 2 - 3 | 407 | 84 | |
BC017049.1 | - | 2 - 3 | 412 | 84 | |
DA890369.1 | - | 2 - 3 | 441 | 84 |