The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014438.4 | NP_055253.2 | 2 - 3 | 117 | 78 |
NM_173178.2 | NP_775270.1 | 2 - 3 | 117 | 78 | |
XM_011510962.1 | XP_011509264.1 | 2 - 3 | 266 | 78 | |
GenBank mRNA | AF200494.1 | - | 2 - 3 | 120 | 78 |
AF201833.1 | - | 2 - 3 | 120 | 78 | |
BC101831.1 | - | 1 - 2 | 71 | 78 | |
BC101833.1 | - | 1 - 2 | 71 | 78 |