The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153027.1 | NP_694572.1 | 1 - 1 | 1594 | 145 |
GenBank mRNA | AK056221.1 | - | 1 - 1 | 1594 | 145 |
BC022381.1 | - | 1 - 1 | 1562 | 145 |