Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24172159-24172432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
UniGene
Hs.460355
Gene Chromosome Location
Chr.16: 23835979-24220611 on Build GRCh38
Transcripts
NM_002738.6 NM_212535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6755772 c.1278C>T p.G426G Chr.16: 24172308-24172308 on Build GRCh38
5508167 c.1316A>T p.K439M Chr.16: 24172346-24172346 on Build GRCh38
5706069 c.1280A>G p.D427G Chr.16: 24172310-24172310 on Build GRCh38
7125939 c.1269G>C p.V423V Chr.16: 24172299-24172299 on Build GRCh38
5706066 c.1280A>C p.D427A Chr.16: 24172310-24172310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750515509 Chr.16: 24172340-24172340 on Build GRCh38
rs752592117 Chr.16: 24172395-24172396 on Build GRCh38
rs192895562 Chr.16: 24172233-24172233 on Build GRCh38
rs772693473 Chr.16: 24172225-24172225 on Build GRCh38
rs747525713 Chr.16: 24172389-24172389 on Build GRCh38
rs989971389 Chr.16: 24172331-24172331 on Build GRCh38
rs769091210 Chr.16: 24172394-24172394 on Build GRCh38
rs761940404 Chr.16: 24172396-24172397 on Build GRCh38
rs754699676 Chr.16: 24172293-24172293 on Build GRCh38
rs536103282 Chr.16: 24172326-24172326 on Build GRCh38
rs555825233 Chr.16: 24172408-24172408 on Build GRCh38
rs898731321 Chr.16: 24172329-24172329 on Build GRCh38
rs772604419 Chr.16: 24172220-24172220 on Build GRCh38
rs868599825 Chr.16: 24172347-24172347 on Build GRCh38
rs769158935 Chr.16: 24172230-24172230 on Build GRCh38
rs758468877 Chr.16: 24172354-24172354 on Build GRCh38
rs377274777 Chr.16: 24172242-24172242 on Build GRCh38
rs369762663 Chr.16: 24172335-24172335 on Build GRCh38
rs978539712 Chr.16: 24172237-24172237 on Build GRCh38
rs776209627 Chr.16: 24172222-24172222 on Build GRCh38
rs751510201 Chr.16: 24172261-24172261 on Build GRCh38
rs767358252 Chr.16: 24172303-24172303 on Build GRCh38
rs764843137 Chr.16: 24172241-24172241 on Build GRCh38
rs748498089 Chr.16: 24172403-24172403 on Build GRCh38
rs761577207 Chr.16: 24172234-24172234 on Build GRCh38
rs762436926 Chr.16: 24172243-24172243 on Build GRCh38
rs766326417 Chr.16: 24172258-24172258 on Build GRCh38
rs754012268 Chr.16: 24172305-24172305 on Build GRCh38
rs780652979 Chr.16: 24172356-24172356 on Build GRCh38
rs747368009 Chr.16: 24172226-24172226 on Build GRCh38
rs1016918004 Chr.16: 24172274-24172274 on Build GRCh38
rs55942437 Chr.16: 24172296-24172296 on Build GRCh38

View Full Product Details